Synura
Synura
Synura, for 30 Students
Catalog # 470176-424
Supplier:   Ward's Science
Supplier #:  SYNURA30/35
Synura
Catalog # 470176-424
Supplier:  Ward's Science
Synura
Synura
Synura, for 30 Students
Price
$15.70
Log in for availability
Each
Product Details & Documents
Freshwater; elongated, biflagellate cells that form globular colonies; gelatinous sheath absent.
Live Cultures are supplied with media in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars. Fast delivery is available directly from our laboratory. Please specify a delivery date.
Each culture shipment is prepared in our laboratories and includes the "Working with Algae & Cyanobacteria" manual which contains detailed instructions for subculturing and use.
Canadian Customers:
CFIA and PHAC regulations require customers to submit a completed inspection checklist and signed statement of facilities compliance before receiving certain cultures. A customer service representative will contact you if documentation is required.
Specifications
  • Désignation:
    Synura, for 30 Students