Synedra
Synedra (Small), Basic culture solution, 30, Tube
Catalog # 470176-422
Supplier: Ward's Science
Fournisseur #: SYNED30/35
Jump to:
Product Details & Documents
Freshwater, Unicellular; pennate, with bilateral symmetry. Diatom or basic medium.
Characterized by shells (tests) made of silica, in two parts.
Spécifications
- Désignation:Synedra (Small)
- No. of Students:30
- Packaging:Tube
- Media:Basic culture solution