Symmetrical Round Jaws Buret Clamps
470005-372
- Description:Symmetrical Round Jaws Buret Clamp, Plain Jaws
- Cat. no.:470005-372
- Supplier No.:7-G37A
Hold round objects from 9.5–35mm in diameter with this versatile clamp.
The jaws can be rotated 360° and may be locked in place at any angle. The clamp can be quickly attached to support rods up to 16mm in diameter. Overall length is 16cm.