ContactUsLinkComponent
Swift M3700 + M3800 Series Advanced Microscopes
Swift M3700 + M3800 Series Advanced Microscopes
Supplier:  Swift Instruments
Certificates
undefined
Swift M3700 + M3800 Series Advanced Microscopes
Supplier:  Swift Instruments
Specifications
  • Catalog No:
  • 470308-158
  • 470308-160
  • Magnification:
  • 40x, 100x, 400x, 1000x (Total)
  • 40x, 100x, 400x, 1000x (Total)
  • Condenser:
  • Abbe NA 1.25 condenser with iris diaphragm
  • Abbe NA 1.25 condenser with iris diaphragm