Swift M3 Micro/Macro Microscope
Micro/Macro Forensic Microscope, LED cordless
Catalog # 470018-298
Supplier: Swift Instruments
Supplier #: M3-F
Jump to:
Product Details & Documents
Combine three microscopes into one.
- Magnification range: 10x to 400x
- Adjustable stage positioning
- 3 year, limited warranty
View specimens in both microscopic and macroscopic views through the M3 monocular with magnification ranging from 10x to 400x. Adjustable stage positioning and a wide range of objectives allow you to quickly transition from compound to macros. Because the unit is cordless, this microscope is ideal for taking along on off-site experiments; it's durable metal construction and unique tripod base allow you to remain sturdy, even on rugged outdoor surfaces. An energy-efficient, rechargeable LED light clearly illuminates even the smallest detail. Includes 3 year, limited warranty.
Specifications
- Désignation:Micro/Macro Forensic Microscope
- Illumination:LED cordless
- Cat. No.:470018-298
- Objectives:4×, 10×, 40×