Specifications
- Description:Compound Monocular Microscope
- Cat. no.:470039-612
- Illumination:Cordless LED
- Supplier No.:M2251C
Specifications
About this item
The Swift Budget Compound Microscope is ideal for teaching elementary, middle, and high school microscopy.
- Full-Size: 34 cm
- Designed for Intermediate Level Microscopy
- Rotatable Head
- Limited Lifetime Manufacturer's Warranty
- Objectives: 4xD, 10xD and 40xR
It offers 4xD, 10xD and 40xR objectives for 40x–400x total magnification. It boasts many student-proof features including locked-on eyepieces and stage clips, a slip-clutch to avoid gear damage, a cord holder that helps avoid cord pull-out, a convenient mode of cord storage, a bottom plate that protects student hands from electrical shock, and retractable 40X objectives that protect the delicate lenses.
Other safety and value-added aspects include a separate coarse and fine focus, a simple design that is easy to repair, an illuminator diffuser that helps reduce heat to your slides, and a special gear lubricant that remains stable even during normal temperature fluctuations and more.