Sunprint Kit
Sunprint Kit
Sunprint Kit
Certificates
Sunprint Kit
2 Options Available Below
Product Details & Documents
The Sunprint Kit includes sun sensitive paper to help you make photographic prints using the power of the sun.
To make images, just place an array of interesting objects on a sheet of Sunprint™ paper, expose it to the sun for a few minutes, immerse in water and watch as a permanent image appears.
Delivery information: Includes 12 sheets of sun-sensitive paper (10×10cm), acrylic plate and instructions.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Kit, Sunprint, 12 Sheets, 10 cm x 10 cm each
    Log in for availability
    Log in for availability
    Each
    Each - $19.50
    $19.50
    Each
       $19.50
    Supplier Number: W110A
  • Refill, Sunprint Kit, 12 Sheets, 10 cm x 10 cm each
    Log in for availability
    Log in for availability
    Pack of 12
    Pack of 12 - $7.40
    $7.40
    Pack of 12
       $7.40
    Supplier Number: W220B