Student Lab Tables for 4 Students with Cabinet or Book Compartment, 1.25" ChemGuard® Top
These tables come in a double-faced design and with a center cabinet that has a single cupboard or a combination drawer/cupboard.
- Available with or without book compartments or drawer cabinet
- 1.25" chemical resistant ChemGuard® work surface
Color: Northwood oak/black
Overall dimensions: 66×42×30"
Cabinet dimensions: 24×40×29"
Drawer dimensions: 11.5×18×5.5"
Frequently Bought Together