Student Filter Paper
Student Filter Paper
Certificates
Student Filter Paper
Student Filter Paper
4 Options Available Below
Product Details & Documents
A good quality paper recommended for student use and all routine filtering operations. This is a strong filter paper with a medium flow rate and retention.
  • Low ash content 0.06%
  • Consistent performance
  • Custom cut
Grade 613 is commonly used in student laboratories. This strong general-purpose filter is also useful in Buchner funnels for barium carbonate, lead chromate and beryllium hydroxides.
Product Family Options
Informations sur le produit
  • Désignation
    Grade
    Ø
    Matière
    Taille de pore
    Availability
    Price
  • Paper, filter, medium flow
    613
    25 mm
    Paper
    6 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $61.20
    $61.20
    Pack of 100
       $61.20
    Supplier Number: 6130-2500
  • Paper, filter, medium flow
    613
    18.5 cm
    Paper
    6 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $57.05
    $57.05
    Box of 100
       $57.05
    Supplier Number: 6130-1850
  • Paper, filter, medium flow
    613
    20 cm
    Paper
    6 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $63.35
    $63.35
    Pack of 100
       $63.35
    Supplier Number: 6130-2000
  • Paper, filter, medium flow
    613
    25 cm
    Paper
    6 µm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $66.60
    $66.60
    Log in pour la disponibilité
    Case of 5000
    Case of 5000 - $2,423.30
    $2,423.30
    Box of 100
       $66.60
    Case of 5000
       $2,423.30
    Supplier Number: 6130-2500