CENCO® String Vibrator | Avantor
CENCO® String Vibrator
String Vibrator
Catalog # 470137-886
Supplier:   Wards
Fournisseur #:  STRVB12
CENCO® String Vibrator
Catalog # 470137-886
Supplier:  Wards
CENCO® String Vibrator
CENCO® String Vibrator
String Vibrator
Price
$256.75
Log in pour la disponibilité
Each
Product Details & Documents
Unit consists of a sturdy metal frame carrying a DC motor with an eccentric shaft for exciting the vibrations. Clamp the frame to a table, attach a string and then simply use masses and a pulley to put tension on the string.
Create standing waves of various frequencies by adjusting the voltage supplied to the motor. Requires a variable-output DC power supply, clamps, slotted masses and a table mounted pulley, not included.
Spécifications
  • Désignation:
    String Vibrator
Accessories
Informations sur le produit
  • Catalog #
    Includes
    Availability
    Price
  • Mass Set of 8, Labware
    Mass Set of 8, Labware
    470017-100
    (1) 10 g, (2) 20 g, (1) 50 g, (1) 100 g, (2) 200 g, (1) 500 g
    Log in pour la disponibilité
    Each - $239.85
Informations sur le produit
  • Catalog #
    Weight
    Availability
    Price
  • Mass Hanger, 500 g Capacity
    Mass Hanger, 500 g Capacity
    470016-120
    5 g
    Log in pour la disponibilité
    Each - $34.75
Informations sur le produit
  • Catalog #
    Ø
    Availability
    Price
  • Clamp Pulley
    Clamp Pulley
    470148-858
    2"
    Log in pour la disponibilité
    Each - $32.20