Streptococcus pneumoniae Slide
Streptococcus pneumoniae Slide, Smear, Gram (+)
Catalog # 470177-408
Supplier: Avantor
Jump to:
Product Details & Documents
Human pathogenic bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Oval to spherical pairs. Agent of pneumonia and meningitis in humans.
Spécifications
- Désignation:Streptococcus pneumoniae Slide
- Stain:Gram (+)
- Orientation:Smear