Streak Plates | Avantor
Streak Plates
Streak Plates
Certificates
Streak Plates
2 Options Available Below
Product Details & Documents
Streak color helps identify minerals.
  • Plates Sized for Field and Lab Testing
  • Unglazed Porcelain
  • Black Plates Work with Light Minerals
These unglazed porcelain plates are sized for field and lab testing. Rubbing the specimen across the plate reveals its streak color.
Black plates work with light minerals.
Product Family Options
Informations sur le produit
  • Désignation
    L×l
    Availability
    Price
  • Streak Plates, White - 10/pk
    2x2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $23.79
    $23.79
    Pack of 10
       $23.79
    Supplier Number: B19183-SP
  • Streak Plates, Black - 10/PK
    2x2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $18.30
    $18.30
    Pack of 10
       $18.30
    Supplier Number: B19182-SP