Epithelium, Composite Slide
Epithelium, Composite Slide, Section, Hematoxylin and Eosin
Catalog # 470177-560
Supplier: Avantor
Jump to:
Product Details & Documents
Compare and contrast epithelium types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Sections of stratified squamous, columnar, and cuboidal epithelium on the same slide.
Specifications
- Désignation:Epithelium, Composite Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section