Fundamentals of Physics Stopped Pendulum
Catalog # 470017-112
Supplier: Wards
Specifications
- Description:FUNdamentals of physics stopped pendulum add-on kit
- Cat. no.:470017-112
- Supplier No.:FP-DP10
Specifications
About this item
Investigate the effect of a stop on a pendulum's period and displacement.
Adjust the pendulum stop to any position in the x- and y-axes
- Hardware mounts precisely to FUN Stand, providing an easy and reliable pendulum stop
Investigate the period and conservation of energy with this great accompaniment to the Fundamentals of Physics Pendulum. The stopped pendulum changes the length of the pendulum mid swing and although the length of the period changes during its motion, the maximum height reached constantly stays the same.
Accessories information:
FUN Stand and FUNdamentals of Phyiscs Pendulum kit are required, but available separately.
Delivery information:
Kit Includes: a pendulum stop, mounting bolt. and instructions.