Sterilizing Tote Boxes
Sterilizing Tote Boxes, 19" x 10 1/2" x 5 1/8"
Catalog # 470173-916
Supplier: Dynalon
Fournisseur #: 410545
Jump to:
Product Details & Documents
This lighter-weight polypropylene tote is easier to carry and can even be autoclaved. Chemically resistant to acids, alkalis, oils, and detergents, it will not rust or corrode. They can be nested for compact storage.
Spécifications
- Désignation:Sterilizing Tote Boxes, 19" x 10 1/2" x 5 1/8"