Ward's® Live Staphylococcus saprophyticus Culture
À propos de cet article
Grows on dead tissues. Novobiocin resistant (S. epidermidis and S. aureus novobiocin sensitive).
- Biosafety Level: 1
- Canada: Containment Level 2 - PHAC Licence required
- Coagulase: Negative
- Catalase: Positive
- Motility: Negative
- Morphology: Cocci in clusters
- Gram reaction: Positive
- Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Grows on dead tissues. Novobiocin resistant (S. epidermidis and S. aureus novobiocin sensitive).
- Biosafety Level: 1
- Canada: Containment Level 2 - PHAC Licence required
- Coagulase: Negative
- Catalase: Positive
- Motility: Negative
- Morphology: Cocci in clusters
- Gram reaction: Positive
- Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Product Family Options
Informations sur le produit
- DésignationMediaIncubation ConditionsStorage ConditionsPackagingAvailabilityPrice
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
- S. saprophyticus : Freeze dried with media packTryptic soy agar37 °CFreezerLyophilizedLog in pour la disponibilitéEach$55.30L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications: