Staphylococcus epidermidis Slide
Catalogue # 470177-412
Fournisseur: Avantor
Spécifications
- Description:Staphylococcus epidermidis Slide
- Cat. no.:470177-412
- Stain:Gram (+)
- Supplier No.:WARD470177-412
- Orientation:Smear
Spécifications
À propos de cet article
Common bacteria on skin
- Identifying features clearly distinguishable
- Rigorous quality control standards
Spheres occurring singly. Pathogenic on human skin.