Boreal Standard Compound Microscopes
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
- Coated-glass optics
- Wide-field eyepiece with pointer
- Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordless charging.
: Microscopes include a manual, dust cover, and limited warranty.
Boreal standard microscopes feature metal construction, a rugged student-proof design, and precision optics making them a favorite among educators.
- Coated-glass optics
- Wide-field eyepiece with pointer
- Limited lifetime warranty
These microscopes optics are parfocal and parcentered for a crisp, consistent field of view. All units feature strategically located controls, course and fine focus, and a 360° rotatable head. Units are available in a variety of configurations offering three or four objectives, plain or mechanical stage, and corded or cordless charging.
: Microscopes include a manual, dust cover, and limited warranty.
- Catalog No:
- 470145-560
- 470145-590
- 470177-090
- 470135-366
- 470145-716
- 470014-588
- 470177-148
- 470177-238
- 470145-732
- 470145-714
- Objectives:
- 4×, 10×, 40×
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×
- 4×, 10×, 40×
- 4×, 10×, 40×
- 4×, 10×, 40×
- 4×, 10×, 40×
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×
Product Family Options
Dual view
- DésignationCondenser TypeIlluminationStage
- : 5745000
Specifications
Cat. No.470145-560Objectives4×, 10×, 40× - : 5584400
Specifications
Cat. No.470145-590Objectives4×, 10×, 40×, 100×
Monocular
- DésignationCondenser TypeIlluminationStage
- : 5745200
Specifications
Cat. No.470177-090Objectives4×, 10×, 40× - : SFC-111LED
Specifications
Cat. No.470135-366Objectives4×, 10×, 40× - : 5584123
Specifications
Cat. No.470145-716Objectives4×, 10×, 40× - : MSH008
Specifications
Cat. No.470014-588Objectives4×, 10×, 40× - : 5584135
Specifications
Cat. No.470177-148Objectives4×, 10×, 40× - : 5584500
Specifications
Cat. No.470177-238Objectives4×, 10×, 40×, 100× - : 5791102
Specifications
Cat. No.470145-732Objectives4×, 10×, 40×, 100× - Compound microscope—TungstenPlain-: -
Specifications
Cat. No.470145-714Objectives4×, 10×, 40×
Substage illuminator for standard microscope
- Désignation
- Illuminator, Substage, for Boreal Standard MicroscopeEach$118.50
Replacement bulb for microscope
- DésignationElectrical