Fixed Blade Scalpel With Fluted Handle | Avantor
Fixed Blade Scalpel With Fluted Handle
Supplier:   Dunrite Instruments
Fixed Blade Scalpel With Fluted Handle
Certificates
Fixed Blade Scalpel With Fluted Handle
Fixed Blade Scalpel With Fluted Handle
1 Options Available Below
Product Details & Documents
Basic scalpels for everyday use.
  • Fixed blades are less likely to shatter
  • Heavy blades are more versatile
  • Easy to sharpen stainless blade
The one-piece design of forged stainless steel provides long-lasting service without the need to replace blades. It features a comfortable fluted handle with a wide 1¹/₂" blade.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Matière
    Longueur
    Désignation
    Availability
    Price
  • Stainless steel
    152 mm (6")
    Stainless Steel Scalpel With Fluted Handle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.30
    $11.30
    Each
       $11.30
    Supplier Number: WLS26715-C
  • Discontinued
    Supplier Number: -