ContactUsLinkComponent
Stainless Steel English and Metric Vernier Caliper
Catalogue # 470110-400
Fournisseur:  CHICAGO BRAND CP
undefined
Stainless Steel English and Metric Vernier Caliper
Catalogue # 470110-400
Fournisseur:  CHICAGO BRAND CP
Supplier Number:  50011
Spécifications
  • Description:
    Vernier Caliper
  • Cat. no.:
    470110-400
  • Measurement Range:
    0 - 150 mm / 0 - 6"
  • Readability:
    0.02 mm / 0.001"
  • Supplier No.:
    50011
Spécifications
À propos de cet article
Dual Scale.
  • Rugged and precise
  • Storage case included