Medium Tip dissecting Forceps | Avantor
Medium Tip dissecting Forceps
Supplier:   Dunrite Instruments
Medium Tip dissecting Forceps
Certificates
Medium Tip dissecting Forceps
Medium Tip dissecting Forceps
2 Options Available Below
Product Details & Documents
Easily Manipulate Tissue Without Sacrificing Hold
  • Available In Curved Or Straight Tip
  • Textured Tip Resists Slipping
  • Stainless Construction Resists Rust
These medium-point forceps have serrated tips and are versatile enough for a variety of dissecting procedures. Some find that curved tips are easier to use. Length: 4 1/2".
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Medium Tip Dissecting Forceps
    Stainless steel
    110 mm (4.5")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.20
    $5.20
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $251.15
    $251.15
    Each
       $5.20
    Box of 50
       $251.15
    Supplier Number: 141001
  • Medium Tip Dissecting Forceps
    Stainless steel
    110 mm (4.5")
    Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7.05
    $7.05
    Each
       $7.05
    Supplier Number: 141003