Broad Tip Dissecting Forceps
Broad Tip Dissecting Forceps
Supplier:   Dunrite Instruments
Broad Tip Dissecting Forceps
Certificates
Broad Tip  Dissecting Forceps
Broad Tip Dissecting Forceps
5 Options Available Below
Product Details & Documents
Easily hold large sections of tissue.
  • Textured tip for a no slip grip
  • Available in multiple sizes
  • Stainless construction resists rust
Broad-pointed, stainless steel forceps with serrated tips are ideal for coarser dissection work such as handling organs and muscle groups. Large sizes are useful for removing specimens from deep jars, etc. They feature a non-slip grip and come in four sizes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Dissecting Forceps
    Stainless steel
    110 mm (4.5")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - 8,75 $
    8,75 $
    Each
       8,75 $
    Supplier Number: 140999
  • Dissecting Forceps
    Stainless steel
    128 mm (5")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 8,00 $
    8,00 $
    Log in for availability
    Box of 50
    Box of 50 - 378,10 $
    378,10 $
    Each
       8,00 $
    Box of 50
       378,10 $
    Supplier Number: 140926
  • Dissecting Forceps
    Stainless steel
    140 mm (5.5")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 9,50 $
    9,50 $
    Log in for availability
    Box of 50
    Box of 50 - 450,60 $
    450,60 $
    Each
       9,50 $
    Box of 50
       450,60 $
    Supplier Number: 141000
  • Dissecting Forceps
    Stainless steel
    203 mm (8")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - 18,95 $
    18,95 $
    Log in for availability
    Box of 25
    Box of 25 - 453,00 $
    453,00 $
    Each
       18,95 $
    Box of 25
       453,00 $
    Supplier Number: 141005
  • Dissecting Forceps
    Stainless steel
    304 mm (12")
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - 26,85 $
    26,85 $
    Each
       26,85 $
    Supplier Number: 141007