Sta-Clear Lens Cleaning Station, Sellstrom
Catalogue # 470006-752
Fournisseur: Sellstrom
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Sta-clear lens cleaning station
- Cat. no.:470006-752
- Supplier No.:23470
Spécifications
À propos de cet article
These water-activated cleansing tissues are safe for glass or plastic lenses and contain an anti-fogging agent.
- Wipe away grime in seconds
The enamel-finished, heavy-gauge metal Sta-Clear cleaning station cabinet easily mounts on the wall and contains a plastic water bottle and pump, a dispenser with 1000 Sta-Clear tissues, and a tissue disposal compartment.
Delivery: Cabinet is supplied complete with key.