Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Stackable Desiccator Cabinet | Avantor

Stackable Desiccator Cabinet

Stackable Desiccator Cabinet
Catalog # 470199-836
Supplier:   Bel-Art
Fournisseur #:  H42053-0000_SE
Stackable Desiccator Cabinet
Catalog # 470199-836
Supplier:  Bel-Art
Stackable Desiccator Cabinet
Stackable Desiccator Cabinet

Stackable Desiccator Cabinet

Price
$606.00
Log in pour la disponibilité
Each
Product Details & Documents

The foam gasket plus two roller latches in the door maintain a tight seal for proper drying conditions and the clear door disappears neatly under the unit when opened.

  • Size: 9.25" L x 8.875" W x 6.625" H
  • Inner dimensions: 8.25" L x 7" W x 5" H

Strong high-impact polystyrene allows you to stack up to five cabinets. A jar of silica gel, for moisture absorption, is included.

Spécifications
  • Désignation:
    Stackable Desiccator Cabinet
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.