Springtail Slide
Springtail, Whole mount, Unstained
Catalog # 470181-608
Supplier: Avantor
Jump to:
Product Details & Documents
Usually Feed on Decaying Plant Matter
- Identifying features clearly distinguishable
- Rigorous quality control standards
Collembola. Shows the furcula, or jumping organ that characterizes this group. Unstained.
Spécifications
- Désignation:Springtail
- Stain:Unstained
- Orientation:Whole mount