Spill Control Station
470204-028
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Spill Control Station
- Width:15"
- Depth:9.5"
- Height:30.75"
- Cat. no.:470204-028
- Supplier No.:68000
Materials to clean up virtually all spills!
- Wall mounted cabinet for easy accessibility
This boldly labeled, wall-mounted white metal cabinet station contains items to clean up virtually all acids, caustic, solvent, or oil spills. It includes ten standard and two hydrofluoric spill control pillows, two pairs of chemical splash goggles, 15 absorbent pads, 15 hazardous waste bags, two pairs of gloves and two total-body coveralls.