Sperm, Rat Slide
Sperm, Rat, Smear, Iron Hematoxylin
Catalog # 470007-806
Supplier: Avantor
Jump to:
Product Details & Documents
Mammal Reproductive System
- Identifying features clearly distinguishable
- Rigorous quality control standards
General structure of mammal sperm.
Specifications
- Désignation:Sperm, Rat
- Stain:Iron Hematoxylin
- Orientation:Smear