Speed Gun
Speed Gun
Catalog # 470039-956
Supplier: BUSHNELL CORPORATION
Fournisseur #: 101911
Jump to:
Product Details & Documents
Simple, Fast, and Fun
- Easy point-and-shoot operation
- A large, clear LCD display
Students are fascinated with speed. Unfortunately it is often tough to measure. This unit uses Bushnell Velocity to measure speed. This device is the ideal tool for tracking everything from racecars to pitching speeds — track the speed of a ball from 100 feet or a car up to 200 mph.
Baseball/softball/tennis: 6–110 mph
Auto racing 6–200 mph
Accuracy: ± 1.0 mph
Spécifications
- Désignation:Speed Gun