Specimen Collecting Kit
Process Any Sample Like the Professionals Do
- Complete Materials for Evidence Collecting
- Easy for Organizing Materials
The kit contains sterile cotton swabs, swab boxes, swab integrity seals, sterile water ampoules, evidence seal, evidence label, biohazard label, pair of nitrile gloves, and antimicrobial hand wipes.
Frequently Bought Together