ContactUsLinkComponent
Southern Blot Analysis Lab Activity
Catalog # 470024-522
Supplier:  Edvotek
undefined
Southern Blot Analysis Lab Activity
Catalog # 470024-522
Supplier:  Edvotek
Supplier Number:  207
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Southern Blot Analysis Lab Activity
  • Cat. no.:
    470024-522
  • Supplier No.:
    207
Specifications
About this item
Lead Students In a Hypothetical Paternity Determination
Introduce students to the procedures of gel-transfer hybridization, also known as Southern Blotting. Students use this technique to detect specific DNA fragments in a simulated paternity determination. DNA fragments are separated by agarose gel electrophoresis, transferred to a membrane and then visualized by staining. This experiment is safe and easy to perform, and does not use human DNA. Includes materials for five blots. Requires agarose electrophoresis chamber, power supply, and 10–20μL micropipets, available separately.