Somso® Synapse Model | Avantor
Somso® Synapse Model
Synapse, 19, 1, 22 cm, 22 cm, 20 cm
Catalog # 470004-080
Supplier:   MARCUS SOMMER
Fournisseur #:  BS 35/3
Somso® Synapse Model
Catalog # 470004-080
Supplier:  MARCUS SOMMER
Somso® Synapse Model
Somso® Synapse Model
Synapse, 19, 1, 22 cm, 22 cm, 20 cm
Price
$821.00
Log in pour la disponibilité
Each
Product Details & Documents
Nerve Components Enlarged 120,000X
  • Highly Detailed Section Of Axon Terminal
  • Shows Details Of Membrane Structure And Neurotransmitter Release
  • 5 Year Manufacturers Warranty
Based on the latest electron microscope interpretation, the model shows all the systems that play a part in the transmission of nerve impulse: neuro tubules and filaments, synaptic vesicles, and membrane structure. The model is mounted on a base and can be removed for closer examination.
Spécifications
  • Désignation:
    Synapse
  • Hauteur:
    20 cm
  • Longueur:
    22 cm
  • No. of Keyed Items:
    19
  • Width:
    22 cm
  • No. of Parts:
    1