ContactUsLinkComponent
Solar System Felt Pieces
Catalogue # 470004-862
Fournisseur:  LITTLE FOLKS VISUALS
undefined
Solar System Felt Pieces
Catalogue # 470004-862
Fournisseur:  LITTLE FOLKS VISUALS
Supplier Number:  4012
Spécifications
  • Description:
    Felt Display Board, 81 cm x 121.9 cm
  • Cat. no.:
    470004-862
  • Supplier No.:
    4012
Spécifications
À propos de cet article
Students have fun manipulating these large brightly colored felt pieces. The solar system has never felt closer.
Caution: WARNING: CHOKING HAZARD - small parts. Not for children under 3 years.