Solar Cells | Avantor
Solar Cells
Solar Cells, 3¾", 2½"
Catalog # 470111-136
Supplier:   Solar Made
Fournisseur #:  4-1.5-200
Solar Cells
Catalog # 470111-136
Supplier:  Solar Made
Solar Cells
Solar Cells
Solar Cells, 3¾", 2½"
Price
$50.45
Log in pour la disponibilité
Each
Product Details & Documents
Show How Direct/Indirect Sunlight Provides Power
Safe, simple-to-use encapsulated mini-panels utilize solar cells similar to those that power satellites in space.
Sturdy panels will endure handling without the normal breakage and cell damage associated with solar cells. A unique lens pattern allows light to be collected from all angles.
The 6" red/black lead wires are attached.
Spécifications
  • Désignation:
    Solar Cells
  • Longueur:
    3¾"
  • Width:
    2½"
  • Cat. No.:
    470111-136
  • Output:
    1.5 V, 200 mA