Solar Cell Unit
Solar Cells, 5", 5"
Catalog # 470016-014
Supplier: Wards
Fournisseur #: PH1320
Jump to:
Product Details & Documents
Show How Direct/Indirect Sunlight Provides Power
Capable of running a small propeller, this solar cell comes mounted on a mini motor. Use this unit to demonstrate the conversion of solar energy into mechanical energy.
Spécifications
- Désignation:Solar Cells
- Longueur:5"
- Width:5"