Soil Sampler with Foot Step
Soil Sampler with Foot Step
Catalog # 470018-632
Supplier: OAKFIELD APPARATUS CO.
Fournisseur #: MODEL K
Jump to:
Product Details & Documents
Replaceable Tip Ensures Long-Lasting Wear
Simply put pressure on the sturdy foot step to force the probe to the desired depth, twist, and remove. It is made from high-quality, chrome-plated steel with a hardened, replaceable tip and includes a cleaning brush and instructions. Overall length: 3'; soil tube length: 11"; tube dia.; 3/4"; weight: 6 lbs.
Spécifications
- Désignation:Soil Sampler with Foot Step
- Cat. No.:470018-632