Soil Sampler Set
Soil Sampler Set, 36", 11/4"
Catalog # 470001-822
Supplier: OAKFIELD APPARATUS CO.
Supplier #: MODEL A
Jump to:
Product Details & Documents
Four-piece set of professional equipment.
- Remove your core intact
- Easy to use in the field
- Aligned with GLOBE protocols
The 12" handle, auger, tube, and two 12" extension rods to reach a depth of 3', are all built to high-grade standards. The screw-on tube tip is replaceable to ensure lifetime usage of this handy tool. Weight: 4 lbs., 12 oz.
Specifications
- Désignation:Soil Sampler Set
- Hauteur:36"
- O.D.:11/4"