Specific Gravity Precision Hydrometer
Precision Specific Gravity Hydrometer, 1.000 - 1.070, 0.0005
470204-506
: Bel-Art
Supplier #: B61801-0700_SE
Precision hydrometer features 0.0005 divisions, 325 mm length, and metal ballast.
- Minute divisions for accurate measurements
- Metal ballast
- Individual serial numbers
- Multi-language statement of accuracy
Hydrometers include individual serial numbers and multi-language (English, French, German, Italian, Portuguese, Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DKD/PTB.
- Désignation:Precision Specific Gravity Hydrometer
- Range:1.000 - 1.070
- Subdivisions:0.0005
- Cat. No.:470204-506
- Accuracy:0.001
- Length:325 mm