Snap Cap Vials
Vials, 4 drams, 30×40 mm, Oui, Snap cap
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 225536
Jump to:
Product Details & Documents
Snap cap vials included tight-fitting polyethylene cap easily snaps off, yet provides a leakproof, evaporation-proof seal.
- Soda lime glass vials
Packaging: Packages of 12.
Specifications
- Avec bouchon:Oui
- Cap Type:Snap cap
- Désignation:Vials, 4 drams
- Diameter ×H:30×40 mm