Sliding Microtome | Avantor
Sliding Microtome
Supplier:   Uchida Yoko
Sliding Microtome
Supplier:  Uchida Yoko
Certificates
Sliding Microtome
Sliding Microtome
2 Options Available Below
Product Details & Documents
Hands-on tool for scientific studies
  • Built in safety features
  • Adjustable section thickness
Cut sections to 40-50 microns safely and easily with this microtome, which features a clear plastic safety cover, fixed blade and rugged plastic construction for hands-on student use.
Simply secure the specimen in the holder, advance using the feed screw, then draw it over the blade. The feed screw moves the specimen only 0.5 mm per complete rotation, and can be advanced as little as 10 microns. For very thin sections, the specimen can be wrapped in an artificial pith.
Also Includes:
Three, two-sided replacement blades
Artificial piths
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Microtome, Sliding
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $551.40
    $551.40
    Each
       $551.40
    Supplier Number: 162-3012
  • Replacement Blade for Sliding Microtome, Set/3
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 3
    Pack of 3 - $61.60
    $61.60
    Pack of 3
       $61.60
    Supplier Number: 916-2304