Single Eye Loupe
Single Lens Loupe, 5×
Catalog # 470021-808
Supplier: American Scientific
Fournisseur #: 4402-00
Jump to:
Product Details & Documents
Ideal for Hands-Free Examination of Small Objects
The 5X lens in a plastic frame fits comfortably over one eye, providing magnification when you need it without interrupting your experiment. Focal length: 2 1/2". Size: 1"L x 1 3/8" widest diameter.
Spécifications
- Grossissement:5×
- Désignation:Single Lens Loupe