Sieves
Sifter, Set/4, 12.5cm dia x 5 cm h Mesh Sizes: 10 mm, 5 mm, 3 mm & 1 mm holes, Nesting, with Lid
Catalog # 470100-684
Supplier: Sempco
Fournisseur #: SS-2638
Jump to:
Product Details & Documents
Set of four, 12.5 cm dia. x 5 cm height each, mesh sizes; 10mm, 5mm, 3mm, and 1mm. Sieves nest for easy storage.
Spécifications
- Désignation:Sifter, Set/4, 12.5cm dia x 5 cm h Mesh Sizes: 10 mm, 5 mm, 3 mm & 1 mm holes, Nesting, with Lid