Sickle Cell Anemia
Catalog # 470177-656
Supplier: Avantor
Specifications
- Description:Sickle Cell Anemia, smear
- Cat. no.:470177-656
- Stain:Giemsa
- Supplier No.:WARD470177-656
- Orientation:Smear
Specifications
About this item
Smear, giemsa stain
- All identifying features are clearly distinguishable
- Rigorous quality control standards
Smears of human peripheral blood collected from a patient with Sickle Cell Anemia. Presence of crescent-shaped red blood cells will vary from patient to patient, thus from slide-to-slide.