Shower Tester
Shower Tester, max shower head diameter 8"
Catalog # 470105-050
Supplier: ENCON SAFETY PRODUCTS
Fournisseur #: 02-5700-01
Jump to:
Product Details & Documents
You know the importance of making sure your safety shower is tested regularly, so it's ready to use in the event of an emergency.
Simply attach this lightweight, PVC coated nylon test funnel to the shower head, and the hemmed bottom passes the water directly and neatly into the bucket.
Will accomodate shower heads up to 8" in diameter. (Bucket not included.)
Spécifications
- Désignation:Shower Tester, max shower head diameter 8"