Sharp-Point Surgical Scissors | Avantor
Sharp-Point Surgical Scissors
Supplier:   Dunrite Instruments
Sharp-Point Surgical Scissors
Certificates
Sharp-Point Surgical Scissors
Sharp-Point Surgical Scissors
3 Options Available Below
Product Details & Documents
Start your cuts easily.
  • Dissect without a scalpel
  • Handles rounded for ambidextrous use
  • Available in two sizes
Sharp-pointed, straight surgical scissors allow the user to insert blades into tissue to start the cut. Larger scissors provide greater leverage for tough cutting; shorter scissors for finer work. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Blade Type
    Availability
    Price
  • Sharp-Point Surgical Scissors
    140 mm (5.5")
    Stainless steel
    Sharp/Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13.85
    $13.85
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $329.50
    $329.50
    Each
       $13.85
    Box of 25
       $329.50
    Supplier Number: 140981
  • Sharp-Point Surgical Scissors
    115 mm (4.5")
    Stainless steel
    Sharp/Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12.50
    $12.50
    Each
       $12.50
    Supplier Number: 140980
  • Surgical Scissors
    165 mm (6.5")
    Stainless steel
    Sharp/Sharp
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.35
    $15.35
    Log in pour la disponibilité
    Box of 10
    Box of 10 - $146.45
    $146.45
    Each
       $15.35
    Box of 10
       $146.45
    Supplier Number: 140982
Accessories
Informations sur le produit
  • Catalog #
    Length
    Material
    Availability
    Price
  • Surgical Scissors
    Surgical Scissors
    470018-942
    165 mm (6.5")
    Stainless steel
    Log in pour la disponibilité
    Each - $15.35