Low-Form Evaporating Dish
Supplier: SILICO & CHEMICO PORCELAIN SE
Jump to:
À propos de cet article
High-Quality Labware, Low Prices
Our laboratory porcelain is high-quality, yet affordably priced. This low-form evaporating dish has a pourout. It is glazed entirely on the inside and over the upper section of the outside. Note: Fragile, use care when handling.
2 Options Available Below
Product Details & Documents
High-Quality Labware, Low Prices
Our laboratory porcelain is high-quality, yet affordably priced. This low-form evaporating dish has a pourout. It is glazed entirely on the inside and over the upper section of the outside. Note: Fragile, use care when handling.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Low-Form Evaporating Dish, Size 1, 30 mLEach$27.85Case of 24$598.10Supplier Number: J-281
- Low-Form Evaporating Dish, Size 2, 50 mLEach$27.85Case of 24$626.60Supplier Number: J-282