Serial Number Restoration Poster
Catalog # 470128-040
Supplier: PRECISION FORENSICS TESTING
Specifications
- Description:Serial Number Restoration Poster
- Cat. no.:470128-040
- Supplier No.:PS-01
Specifications
About this item
A Great Addition to Any Forensics Lab
- Size: 24" x 36"
With detailed depictions of acidic and magnetic serial number restorations, this visual aid is the perfect addition to any serial number restoration activity kit.