Self-Zeroing Automatic Buret Kit
Self-Zeroing Automatic Buret Kit
Catalog # 470211-802
Supplier: Thermo Fisher Scientific
Fournisseur #: 3645-2550
Jump to:
Product Details & Documents
Nalgene® This kit makes any 25 or 50 mL buret both auto-filling and self-zeroing. With 1000 mL squeeze bottle, 1.2 m of clear flexible tubing and fill tube with adapter. Buret not included.
Spécifications
- Désignation:Self-Zeroing Automatic Buret Kit