LaMotte® Secchi Disk
LaMotte® Secchi Disk
Supplier:   LaMotte
LaMotte® Secchi Disk
Supplier:  LaMotte
Certificates
LaMotte® Secchi Disk
LaMotte® Secchi Disk
3 Options Available Below
Product Details & Documents
This 8", black and white disk is used to determine the turbidity of natural water.
  • Black and White design
  • Sounding lead and line included
  • Function: to determine the turbidity of water
  • Aligned with GLOBE protocols
The disk is raised or lowered several times to determine the average depth at which it disappears from sight.
Ordering information: 470112-626 is complete set, containing a secchi disk and a calibrate line. 470004-364 is the standalone rope, and 470101-430 the standalone disk.
Product Family Options
Product Information
  • Désignation
    O.D.
    Availability
    Price
  • Secchi Disk with Calibrated Line
    7.75"
    Log in for availability
    Log in for availability
    Each
    Each - $147.97
    $147.97
    Each
       $147.97
    Supplier Number: 0171-CL
  • Calibrate Line
    20 m
    Log in for availability
    Log in for availability
    Each
    Each - $152.45
    $152.45
    Each
       $152.45
    Supplier Number: 1064-G
  • Secchi Disk Only
    8"
    Log in for availability
    Log in for availability
    Each
    Each - $74.00
    $74.00
    Each
       $74.00
    Supplier Number: 0171