Seawater Analysis Lab Activity
Seawater Analysis Lab Activity
Seawater Analysis Lab Activity
Catalog # 470007-748
Supplier:   Kemtec
Supplier #:  4-102
Seawater Analysis Lab Activity
Catalog # 470007-748
Supplier:  Kemtec
Seawater Analysis Lab Activity
Seawater Analysis Lab Activity
Seawater Analysis Lab Activity
Price
$1,126.55
Log in for availability
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Take an Ocean Voyage Without Leaving Your Classroom
Students analyze the chloride content of seawater by performing titrations using the precipiometric method. The included samples are from the Atlantic and Pacific Oceans, as well as from the Gulf of Mexico. Water from a local source can be analyzed as well. With the included materials, each titration can be repeated three times. It comes with enough materials for 12 setups, student data sheets, and instructional booklet.
Specifications
  • Désignation:
    Seawater Analysis Lab Activity