Scoop, 250 ml, 6.4 cm Handle
470145-134
: Bel-Art
- Description:Scoop, 250mL, 6.4cm Handle
- Cat. no.:470145-134
- Supplier No.:F36753-000
Rigid and strong scoop made from white polypropylene.
- Flat bottom keeps it upright when filled or empty
- Steam autoclavable at 121ºC (250ºF)
Designed for retrieving samples from a larger source.
Scoop, 250mL, 6.4cm Handle